Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFXN3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549032

Catalog No. PA549032

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Potential iron transporter.
Specifications

Specifications

SFXN3
Polyclonal
PBS with 0.09% sodium azide
Q9BWM7
Sideroflexin-3, SFXN3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
81855
Western Blot
Unconjugated
SFXN3
Liquid
SFXN3
KLH conjugated synthetic peptide between 288-314 amino acids from the C-terminal region of human SFXN3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit