Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFXN5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570804
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
Transports citrate; potential iron transporter. Classified as a mitochondrial tricarboxylate carrier protein that has citrate transport activity.
Specifications
SFXN5 | |
Polyclonal | |
Unconjugated | |
Sfxn5 | |
BBG-TCC; C230001H08Rik; SFXN5; sideroflexin 5; sideroflexin-5; Tricarboxylate carrier BBG-TCC | |
Rabbit | |
Affinity chromatography | |
RUO | |
261737 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8CFD0 | |
Sfxn5 | |
synthetic peptide directed towards the N-terminal region of rat SFXN5. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction