Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGCA Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: ABGE-19 , Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19543
Description
SGCA Monoclonal antibody specifically detects SGCA in Human, Mouse, Rat samples. It is validated for IP, WB-Ce, IHC-P.Specifications
SGCA | |
Monoclonal | |
Unconjugated | |
SGCA | |
Rabbit | |
100 μg | |
6442 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
ABGE-19 | |
sarcoglycan, alpha (50kDa dystrophin-associated glycoprotein) | |
50-DAG, A2, ADL, DAG2, DMDA2, LGMD2D, SCARMD1, adhalin | |
A synthetic peptide corresponding to human SGCA | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction