Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGCD Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548734

Catalog No. PA548734

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Component of the sarcoglycan complex, a subcomplex of the dystrophin-glycoprotein complex which forms a link between the F-actin cytoskeleton and the extracellular matrix.
Specifications

Specifications

SGCD
Polyclonal
PBS with 0.09% sodium azide
P82347
Delta-sarcoglycan, Delta-SG, 35 kDa dystrophin-associated glycoprotein, 35DAG, SGCD
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
24052
Western Blot
Unconjugated
Sgcd
Liquid
SGCD
KLH conjugated synthetic peptide between 232-261 amino acids from the C-terminal region of human SGCD.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit