Learn More
Trajan™ Autosampler Syringes With Removable Needle for Thermo Scientific GCs
For reduced non specific sample interaction and increased analyte recovery
$120.25 - $226.83
Specifications
Connection Type Removable
Type Autosampler Syringe
For Use With (Application) Gas chromatography
For Use With (Equipment) Thermo Scientific GC, CTC PAL RTC (Robotic Tool Change) autosampler
Needle Point Style Cone
View More Specs
Products 11
Catalog Number Mfr. No. Diameter (English) Outer Needle Needle Gauge Length (English) Needle Model Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Outer Needle Needle Gauge Length (English) Needle Model Price Quantity & Availability  
SG000680
View Documents
Trajan™
000680
0.024 in. 23 G 2.24 in. 1BR-RTC/RSH-5.7/0.63C
Each for $203.50
Add to cart
SG000480
View Documents
Trajan™
000480
0.024 in. 23 G 2.24 in. 0.5BR-RTC/RSH-5.7/0.63C
Each for $203.50
Add to cart
SG001871
View Documents
Trajan™
001871
0.024 in. 23 G 2.24 in. 5R-RTC/RSH-5.7/0.63C
Each for $166.27
Add to cart
SG001875
View Documents
Trajan™
001875
0.18 in. 26 G 2.24 in. 5R-RTC/RSH-5.7/0.47C
Each for $166.27
Add to cart
SG001877
View Documents
Trajan™
001877
0.18 in. 26 G 3.34 in. 5R-RTC/RSH-8.5/0.47C
Each for $226.83
Add to cart
SG002872
View Documents
Trajan™
002872
0.24 in. 23 G 2.24 in. 10R-RTC/RSH-GT-5.7/0.63C
Each for $162.07
Add to cart
SG002871
View Documents
Trajan™
002871
0.24 in. 23 G 2.24 in. 10R-RTC/RSH-5.7/0.63C
Each for $120.25
Add to cart
SG002874
View Documents
Trajan™
002874
0.24 in. 23 G 3.34 in. 10R-RTC/RSH-GT-8.5/0.63C
Each for $213.26
Add to cart
SG002876
View Documents
Trajan™
002876
0.18 in. 26 G 2.24 in. 10R-RTC/RSH-GT-5.7/0.47C
Each for $162.07
Add to cart
SG002875
View Documents
Trajan™
002875
0.18 in. 26 G 2.24 in. 10R-RTC/RSH-5.7/0.47C
Each for $128.60
Add to cart
SG002878
View Documents
Trajan™
002878
0.18 in. 26 G 3.34 in. 10R-RTC/RSH-GT-8.5/0.47C
Each for $213.26
Add to cart
Description
Description
  • Borosilicate glass barrel for strength and inertness.
  • Stainless steel needles for inertness, strength and corrosion resistance.
  • Backing behind the scale aids quick and accurate reading of the syringe scale.
  • Metal back flange provides stability and resists fracture.
  • Metal needle cap on syringe barrel protects and improves syringe lifetime.
  • Syringes are individually tested for quality ensuring reliability and conformance.
  • Unique packaging design eliminates possible contamination in the syringe.
Specifications
Specifications
Removable
Gas chromatography
Cone
Autosampler Syringe
Thermo Scientific GC, CTC PAL RTC (Robotic Tool Change) autosampler