Learn More
SGE™ BP1 Capillary Columns
Industry-standard nonpolar phase suitable for all routine analyses
Description
Description
  • 100% dimethylpolysiloxane
  • Extremely low-bleed phase with a high temperature limit of 320/340°C