Learn More
Trajan™ Diamond CTC PAL 10 to 50μL GC Autosampler Syringes
Improves sensitivity of MS Analysis with reduced non specific sample interaction and increased analyte recovery.
$141.50 - $160.01
Specifications
Diameter (English) Outer Needle 0.024 in.
Diameter (Metric) Outer Needle 0.63 mm
Volume (Metric) 10 to 50 µL
For Use With (Equipment) CTC PAL
Needle Gauge 23 G
View More Specs
Products 6
Catalog Number Mfr. No. Diameter (Metric) Inner Needle Description Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Needle Description Price Quantity & Availability  
SG0355510
View Documents
Trajan™
0355510
0.24 mm Needle coating: Ultra Smooth Hydrophilic
Each for $144.20
Add to cart
SG0355512
View Documents
Trajan™
0355512
0.11 mm Needle coating: Ultra Smooth Hydrophilic
Each for $144.20
Add to cart
SG0355520
View Documents
Trajan™
0355520
0.24 mm Needle coating: Ultra Smooth Hydrophobic
Each for $160.01
Add to cart
SG0355522
View Documents
Trajan™
0355522
0.11 mm Needle coating: Ultra Smooth Hydrophobic
Each for $160.01
Add to cart
SG0355530
View Documents
Trajan™
0355530
0.24 mm Needle coating: Ultra Smooth
Each for $141.50
Add to cart
SG0355532
View Documents
Trajan™
0355532
0.11 mm Needle coating: Ultra Smooth
Each for $141.50
Add to cart
Description
Description
  • Ensures carryover is reduced to levels lower even than the best fixed needle syringes available. In the event of needle damage, Diamond MS needles provide a cost effective alternative to purchasing an entire fixed needle syringe.
  • Ultra Smooth needles reduce non specific interaction between your sample and the needle. 
  • Further reduce sample interactions with a hydrophobic or hydrophillic needle coating
Specifications
Specifications
0.024 in.
10 to 50 µL
23 G
Cone
0.63 mm
CTC PAL
1.96 in.