Learn More
Trajan™ Diamond GC Needles for 10 to 50μL Manual Syringes
Removable needle design ensures carryover levels are reduced even lower than the best fixed needle syringes available. Trajan Scientific and Medical™ Diamond GC Needles for 10 to 50μL Manual Syringes are MS ready and designed to improve the sensitivity of your MS results.
$133.00 - $161.40
Specifications
Needle Point Style Bevel
For Use With (Equipment) For 10 to 50μL manual syringes
View More Specs
Products 6
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Needle Gauge Description Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Needle Gauge Description Price Quantity & Availability  
SG0355311
View Documents
Trajan™
0355311
0.2 mm 0.5 mm 25 G With Ultra Smooth Hydrophilic coating
Each for $145.25
Add to cart
SG0355310
View Documents
Trajan™
0355310
0.11 mm 0.45 mm 26 G With Ultra Smooth Hydrophilic coating
Each for $150.50
Add to cart
SG0355321
View Documents
Trajan™
0355321
0.2 mm 0.5 mm 25 G With Ultra Smooth Hydrophobic coating
Each for $161.40
Add to cart
SG0355320
View Documents
Trajan™
0355320
0.11 mm 0.45 mm 26 G With Ultra Smooth Hydrophobic coating
Each for $161.40
Add to cart
SG0355330
View Documents
Trajan™
0355330
0.11 mm 0.45 mm 26 G With Ultra Smooth coating
Each for $133.00
Add to cart
SG0355331
View Documents
Trajan™
0355331
0.2 mm 0.5 mm 25 G With Ultra Smooth coating
Each for $133.00
Add to cart
Description
Description
Improve the sensitivity of your MS analysis:
  • Lowest syringe carryover
  • Reduced non-specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Specifications
Bevel
For 10 to 50μL manual syringes