Learn More
Trajan™ Diamond GC Needles for 100 to 500μL Manual Syringes
Removable needle design ensures carryover levels are reduced even lower than the best fixed needle syringes available. Trajan Scientific and Medical™ Diamond GC Needles for 100 to 500μL Manual Syringes are MS ready and designed to improve the sensitivity of your MS results.
$133.00 - $161.40
Specifications
Needle Point Style Bevel
For Use With (Equipment) For 100 to 500μL manual syringes
View More Specs
Products 6
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Needle Gauge Description Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Needle Gauge Description Price Quantity & Availability  
SG0355312
View Documents
Trajan™
0355312
0.2 mm 0.49 mm 25 G With Ultra Smooth Hydrophilic coating
Each for $139.99
Add to cart
SG0355313
View Documents
Trajan™
0355313
0.32 mm 0.6 mm 23 G With Ultra Smooth Hydrophilic coating
Each for $145.00
Add to cart
SG0355323
View Documents
Trajan™
0355323
0.32 mm 0.6 mm 23 G With Ultra Smooth Hydrophobic coating
Each for $161.40
Add to cart
SG0355322
View Documents
Trajan™
0355322
0.2 mm 0.49 mm 25 G With Ultra Smooth Hydrophobic coating
Each for $161.40
Add to cart
SG0355332
View Documents
Trajan™
0355332
0.2 mm 0.49 mm 25 G With Ultra Smooth coating
Each for $133.00
Add to cart
SG0355333
View Documents
Trajan™
0355333
0.32 mm 0.6 mm 23 G With Ultra Smooth coating
Each for $133.00
Add to cart
Description
Description
Improve the sensitivity of your MS analysis:
  • Lowest syringe carryover
  • Reduced non-specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Specifications
Bevel
For 100 to 500μL manual syringes