Learn More
Trajan™ Diamond MS Autosampler Syringes: For Agilent
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
$120.26
Specifications
Connection Type Removable
Type Diamond MS Syringe
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Model Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Model Price Quantity & Availability  
SG002760
View Documents
Trajan™
002760
10 µL 10R-AG-GT-MS1
Each for $120.26
Add to cart
SG003760
View Documents
Trajan™
003760
25 µL 25R-AG-GT-MS1
Each for $120.26
Add to cart
Description
Description
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Specifications
Removable
Diamond MS Syringe