Learn More
Trajan™ Diamond MS Manual Syringes
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
$141.21 - $145.40
Specifications
Connection Type Removable
Type Diamond MS Manual Syringe
Includes Removable ultra smooth needle
View More Specs
Products 6
Catalog Number Mfr. No. Volume (Metric) Model Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Model Price Quantity & Availability  
SG002750
View Documents Promotion Details
Trajan™
002750
10 µL 10R-GT-MS1
Each for $145.40
Add to cart
SG003750
View Documents Promotion Details
Trajan™
003750
25 µL 25R-GT-MS1
Each for $145.39
Add to cart
SG004750
View Documents Promotion Details
Trajan™
004750
50 µL 50R-GT-MS1
Each for $145.39
Add to cart
SG005750
View Documents Promotion Details
Trajan™
005750
100 µL 100R-GT-MS1
Each for $145.39
Add to cart
SG006750
View Documents Promotion Details
Trajan™
006750
250 µL 250R-GT-MS1
Each for $141.21
Add to cart
SG007750
View Documents Promotion Details
Trajan™
007750
500 µL 500R-GT-MS1
Each for $145.39
Add to cart
Description
Description
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Specifications
Removable
Diamond MS Manual Syringe
Removable ultra smooth needle