Learn More
Trajan™ ETP Active Film™ Electron Multipliers
Drop-in, plug-compatible replacement electron multipliers
Supplier: Trajan™ 14566
Description
- Detect presence of ion signals emerging from the mass analyzer of mass spectrometer; also reduce multiplier noise and increase signals
- Available for all major instruments including Agilent Technologies™ Perkin-Elmer™, Shimadzu™, Thermo Scientific, and Varian™; total compatibility with GC/MS, LC/MS, ICP/MS, TOF/MS, and Magnetic Sector techniques
- Features Active Film™—proprietary dynode material with a chemically durable metal oxide secondary emission system; very air stable; exhibits stable performance even after repeated exposure to atmosphere
- Large dynode surface area for extended operating life
- High operational reliability and mechanical robustness; designed for sensitivity and linear system response
- Optimized ion and electron optics for maximum performance
- Easy installation
- Meet EPA, DFTPP ion abundance criteria
- Two-year shelf life
Specifications
Electron Multiplier | |
Varian UltraMass Quadrupole Analyzer Type ICPMS | |
662°F | |
EPA, DFTPP |
ICPMS - Detect Presence of Ion Signals Emerging From The Mass Analyzer of Mass Spectrometer, Also Reduce Multiplier Noise and Increase Signals | |
350°C | |
Large Dynode |
Safety and Handling
ShelfLife : 2 Years