Learn More
SGE™ eVol™ Handheld Automated Analytical Syringe
Improves laboratory workflow
Manufacturer:  SGE™ 2910324
 View more versions of this product
Catalog No. 03-350-291
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.012 in.
0.32mm
For use with eVol automated syringe
50mm
3/Pk.
25 ga.
0.024 in.
0.63mm
1.96 in.
Bevel
eVol analytical syringe
500μL