Learn More
Trajan™ BP10 (1701) GC Capillary Columns
Specifically designed for the separation of organochlorine pesticides listed in EPA methods 608 and 8081
$570.00 - $653.50
Specifications
Coating Fused Silica
Packing Material Forte™ BP10 (1701)
For Use With (Application) Drugs of Abuse, Environmental Analyses (EPA Methods 608 and 8081), Pesticides and Herbicides Analysis, Pharmaceuticals Analysis
Max. Temperature 260/280°C
Polarity Moderately polar
View More Specs
Products 7
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
SG-054253
View Documents
Trajan™
054253
0.22 mm 0.25 µm 25 m Each for $570.00
Add to cart
SG-054268
View Documents
Trajan™
054268
0.32 mm 0.5 µm 25 m Each for $641.50
Add to cart
SG-054280
View Documents
Trajan™
054280
0.53 mm 1 µm 25 m Each for $653.50
Add to cart
SG-054256
View Documents
Trajan™
054256
0.25 mm 0.25 µm 30 m Each for $570.00
Add to cart
SG-054259
View Documents
Trajan™
054259
0.32 mm 0.25 µm 30 m Each for $641.50
Add to cart
SG-054283
View Documents
Trajan™
054283
0.53 mm 1 µm 30 m Each for $653.50
Add to cart
SG-054270
View Documents
Trajan™
054270
0.32 mm 1 µm 30 m This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Highly inert column
  • Low bleed
  • Can be solvent rinsed
  • Ideal for EPA methods 608 and 8081, pesticides, herbicides and pharmaceuticals
Specifications
Specifications
Fused Silica
Drugs of Abuse, Environmental Analyses (EPA Methods 608 and 8081), Pesticides and Herbicides Analysis, Pharmaceuticals Analysis
Moderately polar
G46
Forte™ BP10 (1701)
260/280°C
14% Cyanopropylphenyl Polysiloxane