Learn More
SGE™ BP20 (Wax) GC Capillary Columns
SDP
Combines innovative phase technology and unique fused silica production capability. SGE™ BP20 (Wax) GC Capillary Columns provides end-to-end separation solutions for all applications.
Supplier:  SGE™ 054444
 View more versions of this product
Catalog No. SG-054444
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
Description
Description
Features
  • Industry standard Wax column
  • Bonded and cross-linked
  • Can be solvent rinsed
Applications
  • Used for separation of Alcohol, Free acids, Fatty acids methyl esters, Aromatics, Solvents and Essential oils
Specifications
Specifications
Hydrogen Bonding or Dipole interactions
0.53 mm
260°C
20°C to 240/260°C
30 m
Fused Silica
SGE™ BP20 (Wax)
G14, G15, G16, G20, G39
Free Acids Analysis, Free Acids, Fatty Acid Methyl Esters, Aromatics Analysis, Solvents and Essential Oils
1 μm
Polar
Product Suggestions
Product Suggestions