Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan™ BP21 (FFAP) GC Capillary Columns

A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids

$578.50 - $1064.00

Specifications

Coating Fused Silica
Packing Material BP21 (FFAP)
For Use With (Application) Acrylates, Alcohol Analysis, Aldehydes Analysis, Fatty Acid Methyl Esters, Ketones, Volatile Free Acids
Max. Temperature 250°C
Polarity Polar
View More Specs

Products 10
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
SG-054462
View Documents
Trajan™
054462
0.22 mm 0.25 µm 25 m Each for $578.50
Add to cart
 
SG-054469
View Documents
Trajan™
054469
0.32 mm 0.25 µm 50 m Each for $1,064.00
Add to cart
 
SG-054474
View Documents
Trajan™
054474
0.53 mm 0.5 µm 25 m Each for $690.00
Add to cart
 
SG-054465
View Documents
Trajan™
054465
0.25 mm 0.25 µm 30 m Each for $578.50
Add to cart
 
SG-054466
View Documents
Trajan™
054466
0.25 mm 0.25 µm 60 m Each for $976.00
Add to cart
 
SG-054471
View Documents
Trajan™
054471
0.32 mm 0.25 µm 30 m Each for $628.00
Add to cart
 
SG-054472
View Documents
Trajan™
054472
0.32 mm 0.25 µm 60 m Each for $1,064.00
Add to cart
 
SG-054477
View Documents
Trajan™
054477
0.53 mm 0.5 µm 30 m Each for $690.00
Add to cart
 
SG-054468
View Documents
Trajan™
054468
0.32 mm 0.25 µm 25 m Each for $628.00
Add to cart
 
SG-054478
View Documents
Trajan™
054478
0.53 mm 1 µm 30 m Each for $690.00
Add to cart
 
Description

Description

A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids

  • Nitroterephthalic acid modified PEG polar phase
  • Ideal for low molecular weight acids
  • Applications include volatile free acids, fatty acid methyl esters (FAMEs), alcohols, aldehydes and ketones
Specifications

Specifications

Fused Silica
Acrylates, Alcohol Analysis, Aldehydes Analysis, Fatty Acid Methyl Esters, Ketones, Volatile Free Acids
Polar
G25, G35
BP21 (FFAP)
250°C
Polyethylene Glycol
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit