Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan™ BP21 (FFAP) GC Capillary Columns

A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids

Manufacturer:  Trajan™ 054469

 View more versions of this product

Catalog No. SG-054469


Add to cart

Description

Description

A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids

  • Nitroterephthalic acid modified PEG polar phase
  • Ideal for low molecular weight acids
  • Applications include volatile free acids, fatty acid methyl esters (FAMEs), alcohols, aldehydes and ketones
Specifications

Specifications

Hydrogen Bonding or Dipole interactions
Acrylates, Alcohol Analysis, Aldehydes Analysis, Fatty Acid Methyl Esters, Ketones, Volatile Free Acids
G25, G35
35°C to 240/250°C
50 m
Polar
0.32 mm
Polyethylene Glycol
250°C
0.25 µm
BP21 (FFAP)
Fused Silica
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit