Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trajan™ BP21 (FFAP) GC Capillary Columns
A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids
Supplier: Trajan™ 054471
Description
A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids
- Nitroterephthalic acid modified PEG polar phase
- Ideal for low molecular weight acids
- Applications include volatile free acids, fatty acid methyl esters (FAMEs), alcohols, aldehydes and ketones
Specifications
0.32 mm | |
Hydrogen Bonding or Dipole interactions | |
250°C | |
35°C to 240/250°C | |
30 m | |
Polar |
G25, G35 | |
Polyethylene Glycol | |
Acrylates Analysis, Fatty Acid Methyl Esters, Alcohol Analysis, Aldehydes Analysis, Acrylates, Ketones | |
0.25 μm | |
BP21 (FFAP) | |
Fused Silica |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction