Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trajan BP21 (FFAP) GC Capillary Columns
A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids
Manufacturer: Trajan 054477
Description
A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids
- Nitroterephthalic acid modified PEG polar phase
- Ideal for low molecular weight acids
- Applications include volatile free acids, fatty acid methyl esters (FAMEs), alcohols, aldehydes and ketones
Specifications
Hydrogen Bonding or Dipole interactions | |
Acrylates, Alcohol Analysis, Aldehydes Analysis, Fatty Acid Methyl Esters, Ketones, Volatile Free Acids | |
G25, G35 | |
35°C to 240/250°C | |
30 m | |
Polar |
0.53 mm | |
Polyethylene Glycol | |
250°C | |
0.5 µm | |
BP21 (FFAP) | |
Fused Silica |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title