Learn More
Trajan™ BP5 GC Capillary Column
A general purpose phase used for a wide range of applications
Supplier: Trajan™ 054199
Description
- Excellent general purpose column
- Low bleed
- Non-polar
- High temperature
- Ideal for aromatics, pesticides, herbicides, drugs of abuse and hydrocarbons
Specifications
G36, G41 | |
5% Phenyl/95% Dimethyl Polysiloxane | |
Hydrocarbons Analysis, Pesticide Analysis, Herbicides Analysis, Drugs of Abuse, Hydrocarbons | |
1.5 μm | |
Forte™ BP5 | |
Fused Silica |
0.53 mm | |
320/340°C | |
-60°C to +320/340°C | |
25 m | |
Non polar |