Learn More
Trajan™ BPX5 GC Capillary Columns
MS premium, low bleed phase designed for robustness and is suitable for a wide range of applications
$362.50 - $1251.00
Specifications
Polarity Non polar
Stationary Phase 5% Phenyl Polysilphenylene Siloxane
View More Specs
Products 30
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
SG054099
View Documents
Trajan™
054099
0.1 mm 0.1 µm 10 m
Each for $408.00
Add to cart
SG054100
View Documents
Trajan™
054100
0.25 mm 0.25 µm 15 m
Each for $376.50
Add to cart
SG054112
View Documents
Trajan™
054112
0.22 mm 0.25 µm 12 m
Each for $376.50
Add to cart
SG054114
View Documents
Trajan™
054114
0.22 mm 0.25 µm 50 m
Each for $1,061.00
Add to cart
SG054104
View Documents
Trajan™
054104
0.15 mm 0.25 µm 25 m
Each for $561.00
Add to cart
SG054118
View Documents
Trajan™
054118
0.32 mm 0.25 µm 12 m
Each for $373.50
Add to cart
SG054121
View Documents
Trajan™
054121
0.25 mm 1 µm 15 m
Each for $376.50
Add to cart
SG054122
View Documents
Trajan™
054122
0.25 mm 1 µm 30 m
Each for $668.50
Add to cart
SG054119
View Documents
Trajan™
054119
0.32 mm 0.25 µm 25 m
Each for $658.50
Add to cart
SG054126
View Documents
Trajan™
054126
0.32 mm 0.5 µm 50 m
Each for $1,251.00
Add to cart
SG0541261
View Documents
Trajan™
0541261
0.32 mm 1 µm 6 m
Each for $362.50
Add to cart
SG054125
View Documents
Trajan™
054125
0.32 mm 0.5 µm 25 m
Each for $658.50
Add to cart
SG054127
View Documents
Trajan™
054127
0.32 mm 1 µm 12 m
Each for $373.50
Add to cart
SG054130
View Documents
Trajan™
054130
0.53 mm 1 µm 12 m
Each for $431.00
Add to cart
SG054129
View Documents
Trajan™
054129
0.32 mm 1 µm 50 m
Each for $1,251.00
Add to cart
SG054131
View Documents
Trajan™
054131
0.53 mm 1 µm 25 m
Each for $717.50
Add to cart
SG0541345
View Documents
Trajan™
0541345
0.53 mm 0.5 µm 30 m
Each for $717.50
Add to cart
SG054134
View Documents
Trajan™
054134
0.53 mm 0.25 µm 25 m
Each for $717.50
Add to cart
SG054133
View Documents
Trajan™
054133
0.53 mm 0.25 µm 12 m
Each for $431.00
Add to cart
SG054154
View Documents
Trajan™
054154
0.32 mm 1 µm 60 m
Each for $1,251.00
Add to cart
SG054142
View Documents
Trajan™
054142
0.22 mm 0.25 µm 30 m
Each for $668.50
Add to cart
SG054148
View Documents
Trajan™
054148
0.53 mm 1 µm 30 m
Each for $717.50
Add to cart
SG054146
View Documents
Trajan™
054146
0.32 mm 0.25 µm 60 m
Each for $1,251.00
Add to cart
SG054145
View Documents
Trajan™
054145
0.32 mm 0.25 µm 30 m
Each for $658.50
Add to cart
03-350-357
View Documents
Trajan™
054113
0.22 mm 0.25 µm 25 m
Each for $668.50
Add to cart
03-350-356
View Documents
Trajan™
054102
0.25 mm 0.25 µm 60 m
Each for $1,061.00
Add to cart
SG054144
View Documents
Trajan™
054144
0.25 mm 0.25 µm 15 m
Each for $373.50
Add to cart
SG054110
View Documents
Trajan™
054110
0.15 mm 0.15 µm 30 m
Each for $561.00
Add to cart
SG0541011
View Documents
Trajan™
0541011
0.25 mm 0.1 µm 30 m
Each for $668.50
Add to cart
SG-0542170
View Documents
Trajan™
0542170
0.25 mm 0.1 µm 15 m
Each for $376.50
Add to cart
Description
Description
  • Very low bleed
  • High temperature
  • Non-polar
  • Ideal for GCMS
  • Applications include pesticides, herbicides, hydrocarbons, amines and phenols
Specifications
Specifications
Non polar
5% Phenyl Polysilphenylene Siloxane