Learn More
Trajan™ HT8 GC Capillary Columns
Unique, high temperature phase designed for the analysis of polychlorinated biphenyl (PCB) congeners
$620.00 - $1111.00
Specifications
Coating Fused Silica
Film Thickness 0.25 µm
Packing Material Forte™ HT8
For Use With (Application) Environmental Analysis, Nitro-substituted Aromatics, PCB Congener Analysis, Pesticides and Herbicides Analysis, Polynuclear Aromatic Hydrocarbons
Max. Temperature 360/370°C
View More Specs
Products 5
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Price Quantity & Availability  
SG-054675
View Documents
Trajan™
054675
0.22 mm 25 m Each for $620.00
Add to cart
SG054682
View Documents
Trajan™
054682
0.32 mm 60 m Each for $1,111.00
Add to cart
SG-054680
View Documents
Trajan™
054680
0.32 mm 25 m Each for $642.00
Add to cart
SG054677
View Documents
Trajan™
054677
0.25 mm 30 m Each for $620.00
Add to cart
03-350-364
View Documents
Trajan™
054683
0.25 mm 60 m Each for $1,107.00
Add to cart
Description
Description
  • High temperature, low bleed column
  • Preferred column for polychlorinated biphenyl (PCB) analysis
  • Separates PCBs on ortho ring as well as boiling point
Specifications
Specifications
Fused Silica
Forte™ HT8
360/370°C
0.25 µm
Environmental Analysis, Nitro-substituted Aromatics, PCB Congener Analysis, Pesticides and Herbicides Analysis, Polynuclear Aromatic Hydrocarbons
8% Phenyl (equiv.) Polycarborane Siloxane