Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGE™ MEPS™ BINs

Available in needle styles to suit LC and GC applications

Manufacturer:  SGE™ 2900105

 View more versions of this product

Catalog No. 03-350-139


Add to cart

Description

Description

The MEPS approach to sample preparation is suitable for reversed phases, normal phases, mixed mode and ion exchange chemistries.

  • For GC applications: needle is 23 gauge, 0.63mm OD, cone point style
  • For LC applications: needle is 22 gauge, 0.72mm OD, LC point style
  • Typical BIN life is conservatively about 40-100 samples but actual life is dependant on the sample matrix
  • The dimensions of the sorbent bed ensure that the sorbent performance remains identical to conventional SPE devices used for extraction of similar samples
  • Sample preparation for complex biological samples is readily adapted to MEPS and reduces the volume of sample and reagents required for extraction when compared with conventional SPE and other “micro extraction” procedures
  • Needles are packed in a sealed blister pack of 5 needles to ensure their integrity
  • Suitable for use with eVol™ automated syringes

Specifications

Specifications

Silica
23G
MEPS development kit
MEPS
HTA 300A plus, Thermo Scientific™, Varian 8400 and 100μL CTC analytics system
60Å
One each C18, C8, C2, Silica, C8+SCX
45μm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit