Learn More
Trajan™ MEPS BINs for LC Applications
MEPS is Micro Extraction by Packed Sorbent. Provides miniaturization of conventional sample preparation devices
Manufacturer:  Trajan™ 2900501
 View more versions of this product
Catalog No. 03-350-153
  Check Availability
Add to cart
Description
Description
  • Save hours in sample preparation
  • Reduce solvent consumption and sample volume
  • Available in reversed phase, normal phase, mixed mode and ion exchange chemistries
Specifications
Specifications
LC
Liquid chromatography
22g
MEPS BINs
LC point needle
10μL to 1mL or more
0.72mm
CTC Analytics systems using 250μL syringes
60Å
C18
0.0003 to 0.03 oz. or More
45μm