Learn More
SGE™ MEPS BINs for LC applications
Manufacturer:  SGE™ 2900402
 View more versions of this product
Catalog No. 03-350-148
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
LC
Liquid chromatography
22g
MEPS BINs
LC point needle
10μL to 1mL or more
0.72mm
HTA 300APlus, Thermo Scientific, Varian 8400 and 100μL CTC Analytics systems
60Å
Silica
0.0003 to 0.03 oz. or More
45μm