Learn More
Trajan PEEKsil Tubing
Mechanically robust polymer-sheathed fused silica tubing. Trajan Scientific and Medical PEEKsil Tubing can be used for sealing with conventional metal or polymer ferrule systems.
$82.00 - $134.00
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Diameter (Metric) Outer | Overall Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Diameter (Metric) Outer | Overall Length (Metric) | Price | Quantity & Availability | |||
SG-062451
|
Trajan
0624251 |
0.05 mm | 1.5 mm | 50 mm |
Pack of 5 for $82.00
|
|
SG-0624350
|
Trajan
0624350 |
0.175 mm | 1.5 mm | 500 mm |
Pack of 2 for $99.00
|
|
SG-0624252
|
Trajan
0624252 |
0.05 mm | 1.5 mm | 100 mm |
Pack of 5 for $134.00
|
|
SG-0624351
|
Trajan
0624351 |
0.175 mm | 1.5 mm | 50 mm |
Pack of 5 for $82.00
|
|
SG-0624352
|
Trajan
0624352 |
0.175 mm | 1.5 mm | 100 mm |
Pack of 5 for $134.00
|
|
SG-0624353
|
Trajan
0624353 |
0.175 mm | 1.5 mm | 200 mm |
Pack of 5 for $108.00
|
|
SG-0624250
|
Trajan
0624250 |
0.05 mm | 1.6 mm | 500 mm |
Pack of 2 for $95.00
|
|
SG-0624253
|
Trajan
0624253 |
0.05 mm | 1.5 mm | 200 mm |
Pack of 5 for $108.00
|
|
Description
- Robust - PEEK polymer exterior and fused silica inner tubing
- Smooth wall surface giving excellent flow characteristics
- Compatible with most organic solvents
- Inertness - extremely low absorption characteristics
- Precise ID
Specifications
Tubing |