Learn More
Trajan™ Replacement Needles for Thermo Scientific GC Syringes
Cone tip style. Trajan Scientific and Medical™ Replacement Needles for Thermo Scientific GC Autosampler Syringes are not gas tight.
Manufacturer:  Trajan™ 033770
 View more versions of this product
Catalog No. 03-350-124
  Check Availability
Add to cart
Description
Description
  • Needle length is the exposed length of needle
  • Fits Thermo Scientific™ syringes for AS3000, AS2000 and TriPlus RSH Autosamplers
Specifications
Specifications
26g
0.47mm
50mm
GC Autosampler Syringe Replacement Needle
0.155mm
0.5μL
Cone