Learn More
Trajan™ Replacement Plungers
Replacement Plungers are available for all SGE gas tight syringes.
Manufacturer:  Trajan™ 031816
 View more versions of this product
Catalog No. 03-350-106
  Check Availability
Add to cart
Description
Description
  • For use with SGE gas tight syringes.
  • Use the SGE Syringe Part Number to find a replacement plunger.
Specifications
Specifications
For 25uL fixed needle gas tight syringe for Carlo Erba CTC or Fisons autosamplers; Syringes 003988, 003985
IN VITRO use only. For lab analyzers only. Not for human use.
PTFE tip
Replacement Plunger, P25R-C/T-GT