Learn More
Trajan™ Replacement Plungers for Diamond Manual Syringes
SGE syringe replacement plungers for gas tight syringes
$71.70
Specifications
Material (Plunger) PTFE
Type Replacement Plunger
View More Specs
Products 5
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
SG0355102
View Documents
Trajan™
0355102
25μL Diamond manual syringe
Each for $71.70
Add to cart
SG0355105
View Documents
Trajan™
0355105
50μL Diamond manual syringe
Each for $71.70
Add to cart
SG0355101
View Documents
Trajan™
0355101
10μL Diamond manual syringe
Each for $71.70
Add to cart
SG0355110
View Documents
Trajan™
0355110
100μL Diamond manual syringe
Each for $71.70
Add to cart
SG0355120
View Documents
Trajan™
0355120
250μL Diamond manual syringe
Each for $71.70
Add to cart
Description
Description
  • Replacement plungers for gas tight syringes
  • Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
PTFE
Replacement Plunger