SGE™ SilFlow™ GC Replacement Parts
Tightened using finger force to achieve a reliable seal, even for sensitive MS systems
Manufacturer: SGE™ 123705
 View all options for this product
Catalog No. 03-350-246
  Check Availability
Add to Cart
    Type Ferrules
    Diameter (Metric) Inner 1.1mm ID
    For Use With (Equipment) SGE Silflow microchannel device
    • Conventional graphite or Vespel™ ferrules cannot be used with the SilFlow MCD as the dimensional stability is not adequate and there is a risk of contaminating the channels
    • SilTite™ FingerTite metal ferrules result in a reliable zero dead volume connection
  • Description & Specifications
Related Products
Product Content Feedback