Learn More
SGE™ SilFlow™ GC Replacement Parts
Tightened using finger force to achieve a reliable seal, even for sensitive MS systems
Manufacturer:  SGE™ 123706
 View more versions of this product
Catalog No. 03-350-247
  Check Availability
Add to cart
Description
Description
  • Conventional graphite or Vespel™ ferrules cannot be used with the SilFlow MCD as the dimensional stability is not adequate and there is a risk of contaminating the channels
  • SilTite™ FingerTite metal ferrules result in a reliable zero dead volume connection
Specifications
Specifications
SGE Silflow microchannel device
Ferrule
0.36mm ID