Learn More
Invitrogen™ SGEF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109410
Description
Immunogen sequence: ICQKTPKDSP KYEVCKRALK EVSKLVRLCN EGARKMERTE MMYTINSQLE FKIKPFPLVS SSRWLVKRGE LTAYVEDTVL FSRRTSKQQV YFFLFNDVLI ITKKKSEESY NVNDYSLRDQ LLVESCDNEE LNSSPGKNSS TMLYSRQSSA SHLFTLTVLS NHANEKVEML LGAETQSERA RWITALGHSS GKPPADRTSL TQVEIVRSFT AKQPDELSLQ VADVVLIYQR VSDGWYEGER LRDGERGWFP MECAKEITCQ ATIDKNVERM GRLLGLETNV.
Activates RhoG GTPase by promoting the exchange of GDP by GTP. Required for the formation of membrane ruffles during macropinocytosis. Required for the formation of cup-like structures during trans-endothelial migration of leukocytes. In case of Salmonella enterica infection, activated by SopB, which induces cytoskeleton rearrangements and promotes bacterial entry.
Specifications
SGEF | |
Polyclonal | |
Unconjugated | |
ARHGEF26 | |
4631416L12Rik; 8430436L14Rik; AI303572; ARHGEF26; AW491953; CSGEF; HMFN1864; RGD1562559; Rho guanine nucleotide exchange factor (GEF) 26; Rho guanine nucleotide exchange factor 26; SGEF; SH3 domain-containing guanine exchange factor; Src homology 3 domain-containing guanine nucleotide exchange factor; testicular tissue protein Li 171 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26084, 310460, 622434 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.75 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q96DR7 | |
ARHGEF26 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 592-871 of human ARHGEF26 (NP_056410.3). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.