Learn More
SGK1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 233941AP
Catalog No. 50-173-4067
Add to cart
Description
Description
This antibody is specific to SGK1.
Serum and glucocorticoid-regulated protein kinase 1 (SGK1) is a member of the AGC family of serine/threonine kinases that plays an important role in cellular stress response and cell survival. SGK1 is transcriptionally activated by numerous stimuli, including serum, glucocorticoids, osmotic stress, and hormones. SGK1 functions as a regulator of cell survival and ion channels and transporters such as ENaC or KCNA3/Kv1.3. It phosphorylates and negatively regulates pro-apoptotic FOXO3A. SGK1 is thought to play an essential function in nephropathy and cardiovascular development.
Specifications
Specifications
SGK1
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00141, Q9WVC6
SGK, SGK1
Rabbit
IgG
150 μL
-20°C
Primary
20393, 6446
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
SGK1
Liquid
SGK1
SGK1 Fusion Protein Ag17888
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse