Learn More
SGK196 Mouse anti-Human, Clone: OTI2E6, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF804718
Catalog No. 50-166-7104
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The exact function of FLJ23356 remains unknown.
Specifications
Specifications
SGK196
Monoclonal
Unconjugated
POMK
Lyophilized
POMK
Human recombit protein fragment corresponding to amino acids 141-350 of human SGK196 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI2E6
PBS with 8% trehalose and no preservative; pH 7.3
Q9H5K3
MDDGA12, MDDGC12, SGK196
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
84197