Learn More
SGMS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 190501AP
Catalog No. 50-173-2326
Add to cart
Description
Description
A bidirectional lipid cholinephosphotransferases capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) and the inverse reaction. Direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. Directly and specifically recognizes the choline head group on the substrate. Also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. May not function strictly as a SM synthase. Inhibited by bacterial PC-phospholipase C inhibitor D609. Localized to Golgi apparatus; Golgi membrane; multi-pass membrane protein. Possibly present on Brain, heart, kidney, liver, muscle and stomach.
Specifications
Specifications
SGMS1
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7TSX5, Q86VZ5, Q8VCQ6
hmob33, MOB, MOB1, Protein Mob, SGMS1, SMS1, sphingomyelin synthase 1, TMEM23, Transmembrane protein 23
Rabbit
IgG
150 μL
-20°C
Primary
208449, 259230, 353229
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
SGMS1
Liquid
Sgms1
SGMS1 Fusion Protein Ag5246
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat