Learn More
SGOL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 169771AP
Catalog No. 50-173-1306
Add to cart
Description
Description
The protein encoded by this gene is a member of the shugoshin family of proteins. This protein is thought to protect centromeric cohesin from cleavage during mitotic prophase by preventing phosphorylation of a cohesin subunit. Reduced expression of this gene leads to the premature loss of centromeric cohesion, mis-segregation of sister chromatids, and mitotic arrest. Evidence suggests that this protein also protects a small subset of cohesin found along the length of the chromosome arms during mitotic prophase. An isoform lacking exon 6 has been shown to play a role in the cohesion of centrioles. Mutations in this gene have been associated with Chronic Atrial and Intestinal Dysrhythmia (CAID) syndrome, characterized by the co-occurrence of Sick Sinus Syndrome (SSS) and Chronic Intestinal Pseudo-obstruction (CIPO) within the first four decades of life. Fibroblast cells from CAID patients exhibited both increased cell proliferation and higher rates of senescence. Pseudogenes of this gene have been found on chromosomes 1 and 7. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
SGOL1
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5FBB7, Q9CXH7
hSgo1, NY BR 85, SGO, Sgo1, SGOL1, Shugoshin like 1, shugoshin like 1 (S. pombe)
Rabbit
IgG
150 μL
-20°C
Primary
151648, 363174, 72415
Western Blot
Unconjugated
SGO1
Liquid
Sgo1
SGOL1 Fusion Protein Ag9700
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat