Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse APS, Synthetic Peptide, Invitrogen™

Catalog No. PEP246
Click to view available options
Quantity:
50 μg

Synthetic Peptide

SH2B2/APS Synthetic Peptide, PEP-246, from Invitrogen™.

APS (adapter protein with Pleckstrin homology and Src homology 2 domains) a member of the Lnk family, is an adaptor protein that is involved in B-cell signaling, insulin signaling, cytoskeletal reorganization. It is tyrosine phosphorylated by JAK2, KIT and other kinases during B-cell receptor stimulation of many different cytokines, chemokines and leukokines. APS has been shown to inhibit the JAK-STAT pathway in collaboration with c-Cbl. It is located in the cytoplasm in pre-PDGF (platelet derived growth factor) stimulated cells and localizes to the plasma membrane and peripheral regions upon PDGF stimulation. This protein is expressed in B-cells and mast cells when detected in haematopoietic cell lines yet is also expressed in many different tissues including spleen, prostrate, testis, uterus, small intestine and skeletal muscle. This peptide corresponds to amino acids E(605) ATLGRARAVENQYS (619) from mouse APS and can be used as a blocking peptide with polyclonal antibody PA1-1062.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 23921
Species Mouse
Purification Method purified
Accession Number Q9JID9
Common Name Mouse APS
Concentration 1 mg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Lyophilized
Formulation water with no preservative
For Use With (Application) Blocking Assay, Control
Gene Alias Alpha-1-antiproteinase S, Alpha-1-antitrypsin, Alpha-1-proteinase inhibitor
Gene Symbol Sh2b2
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.