Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH2B3 Mouse anti-Human, Clone: OTI1F6, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503069

Catalog No. 50-167-4043


Add to cart

Description

Description

This gene encodes a member of the SH2B adaptor family of proteins, which are involved in a range of signaling activities by growth factor and cytokine receptors. The encoded protein is a key negative regulator of cytokine signaling and plays a critical role in hematopoiesis. Mutations in this gene have been associated with susceptibility to celiac disease type 13 and susceptibility to insulin-dependent diabetes mellitus.
Specifications

Specifications

SH2B3
Monoclonal
0.43 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UQQ2
IDDM20, LNK
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10019
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1F6
Unconjugated
SH2B3
Liquid
SH2B3
Full length human recombit protein of human SH2B3 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit