Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ SH2D1B Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$280.50 - $1004.00

Specifications

Formulation PBS (pH 7.4) containing 20% glycerol. containingno Preservative
Gene Alias EAT2EAT-2, EWS/FLI1-activated transcript 2, SH2 domain containing 1B, SH2 domain-containing molecule EAT2, SH2 domain-containing protein 1B
Gene ID (Entrez) 117157
Gene Symbol SH2D1B
Name Recombinant SH2D1B Protein
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB60211250U View Documents Novus Biologicals™
NBP260211250UG
250μg Each for $1,004.00
Add to cart
 
NB6021150UG View Documents Novus Biologicals™
NBP26021150UG
50μg Each for $280.50
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-132 of Human SH2D1B The Recombinant Human SH2D1B Protein is derived from E. coli. The Recombinant Human SH2D1B Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

PBS (pH 7.4) containing 20% glycerol. containingno Preservative
117157
Recombinant SH2D1B Protein
Baculovirus
0.25mg/mL
SDS-Page
EAT2EAT-2, EWS/FLI1-activated transcript 2, SH2 domain containing 1B, SH2 domain-containing molecule EAT2, SH2 domain-containing protein 1B
SH2D1B
Recombinant Protein
RUO
< 1.0 EU per 1ug of protein (determined by LAL method)

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit