Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH2D4A Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 159571AP

Catalog No. 50-173-0599


Add to cart

Description

Description

SH2D4A inhibits estrogen-induced cell proliferation by competing with PLCG for binding to ESR1, blocking the effect of estrogen on PLCG and repressing estrogen-induced proliferation. SH2D4A may play a role in T-cell development and function.
Specifications

Specifications

SH2D4A
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H788
Protein SH(2)A, SH2 domain containing 4A, SH2A, SH2D4A
Rabbit
IgG
150 μL
-20°C
Primary
63898
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
SH2D4A
Liquid
SH2D4A
SH2D4A Fusion Protein Ag8713
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit