Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP27 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548390

Catalog No. PA548390

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Rho GTPase-activating protein which may be involved in clathrin-mediated endocytosis. GTPase activators for the Rho-type GTPases act by converting them to an inactive GDP-bound state. Has activity toward CDC42 and RAC1.
Specifications

Specifications

ARHGAP27
Polyclonal
PBS with 0.09% sodium azide
Q6ZUM4
Rho GTPase-activating protein 27, CIN85-associated multi-domain-containing Rho GTPase-activating protein 1, Rho-type GTPase-activating protein 27, SH3 domain-containing protein 20, ARHGAP27, CAMGAP1, SH3D20
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
201176
Western Blot
Unconjugated
ARHGAP27
Liquid
ARHGAP27
KLH conjugated synthetic peptide between 198-226 amino acids from the Central region of human SH3D20.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit