Learn More
Aviva Systems Biology Corp SH3GL2 Antibody - middle region (ARP56550_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56550_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SH3GL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SH3GL2. Alternate names: CNSA2, EEN-B1, FLJ20276, FLJ25015, SH3D2A, SH3P4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61901