Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3GL3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17963020UL

 View more versions of this product

Catalog No. NBP17963020


Add to cart

Description

Description

SH3GL3 Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

SH3GL3
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
SH3GL3
Synthetic peptide directed towards the middle region of human Sh3gl3The immunogen for this antibody is Sh3gl3. Peptide sequence IDPLQLLQDKDLKEIGHHLRKLEGRRLDYDYKKRRVGKIPEEEIRQAVEK.
38 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Western Blot
Western Blot 1:1000
NP_059096
CNSA3, EEN-B2, endophilin-A3, SH3D2C, SH3-domain GRB2-like 3
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
6457
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit