Learn More
SH3GLB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 158971AP
Catalog No. 50-173-0559
Add to cart
Description
Description
Belongs to the endophilin family. 2 isoforms of the human protein are produced by alternative splicing. Pierrat B., Genomics 71:222-234(2001). Nagase T., DNA Res. 8:85-95(2001). Wan D., Proc. Natl. Acad. Sci. U. S. A. 101:15724-15729(2004).
Specifications
Specifications
SH3GLB2
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5PPJ9, Q8R3V5, Q9NR46
Endophilin B2, KIAA1848, PP6569, PP9455, SH3GLB2
Rabbit
IgG
150 μL
-20°C
Primary
227700, 311848, 56904
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
SH3GLB2
Liquid
SH3GLB2
SH3GLB2 Fusion Protein Ag8712
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat