Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3MD2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15497220UL

 View more versions of this product

Catalog No. NBP15497220


Add to cart

Description

Description

SH3MD2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

SH3MD2
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
SH3RF1
Synthetic peptides corresponding to SH3RF1(SH3 domain containing ring finger 1) The peptide sequence was selected from the middle region of SH3RF1. Peptide sequence LLKLLSGASTKRKPRVSPPASPTLEVELGSAELPLQGAVGPELPPGGGHG.
93 kDa
20ul
Zinc Finger
Primary
57630
Western Blot
Western Blot 1:100-1:2000
Q7Z6J0
EC 6.3.2, EC 6.3.2.-, KIAA1494SH3 multiple domains protein 2, Plenty of SH3s, POSHFLJ21602, Protein POSH, putative E3 ubiquitin-protein ligase SH3RF1, RING finger protein 142, RNF142plenty of SH3 domains, SH3 domain containing ring finger 1, SH3 domain-containing RING finger protein 1, SH3 multiple domains 2, SH3MD2
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit