Learn More
Thermo Scientific™ Shandon™ AN-69 Dissecting Tray
Work on an elevated surface with this stainless-steel dissecting tray.
Manufacturer:  Thermo Scientific™ 11469
Catalog No. 11469
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Work on an elevated surface with Thermo Scientific™ Shandon™ AN-69 Dissecting Tray.Thermo Scientific™ Shandon AN-69 Dissecting Tray provides an elevated working surface for dissection.
Features:
  • Heavy-gauge stainless-steel tray accommodates polyethylene dissecting boards.
  • Work surface is cross-creased for positive drainage and has 5 in. (12.7cm) long drain.
  • Perforated grid plate can be removed for easy cleaning.
Specifications
Specifications
12.7cm