Learn More
Thermo Scientific™ Shandon™ AN-70 Mortuary Rack
Store cadavers in refrigerated rooms on sturdy, free-standing Shandon AN-70 Mortuary Racks.
Manufacturer:  Thermo Scientific™ 10360
 View more versions of this product
Catalog No. 23-245-381
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Store cadavers in refrigerated rooms on sturdy, free-standing Thermo Scientific™ Shandon AN-70 Mortuary Racks.
Features:   
  •  Excellent for use in walk-in refrigerators
  •  Heavy gauge Type 304 stainless-steel construction
  •  Adjustable 11-gauge stainless-steel tray supports provide flexibility for your facility
  •  Tray supports can be adjusted to fit your facility's requirements; factory suggested spacing is 18" (45.7cm) increments
  •  Lowest tray support is permanently welded to the frame for additional strength
  •  Top and bottom tray supports are accessible with the AN-30 lift
  •  Units can be free-standing for flexibility or secured to floor for added stability
  •  Units are available to accommodate standard and bariatric body trays and AN-49 transportable autopsy tabletops.
Ordering information:
  • Available in three-, four- and five-tier sizes
Description & Specifications
Specifications
Stainless Steel
Mortuary Racks
41.3 in.
104.9cm
53 in.
134.6cm
70.3 in.
6.9 in.
64.9 in.
Shandon™
4
31 in.
78.7cm
80 in.
203.2cm
178.6cm
17.6cm
164.8cm