Learn More
Thermo Scientific™ Shandon™ Autopsy Headrests
Protect autopsy surfaces from scratches with these headrests.
Manufacturer:  Thermo Scientific™ 408
 View more versions of this product
Catalog No. 408
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Protect autopsy surfaces from scratches with Thermo Scientific™ Shandon™ Autopsy Headrests.
  • Ten positions of height, width, length and curvature vary on the faces of the headrests for maximum accommodation
  • Only four points contact the table, and base is curved to permit drainage materials to flow freely
  • Each side of the unit is deeply curved to hold the head securely
  • Available in autoclavable aluminum or semi-hard rubber
Specifications
Specifications
Aluminum
Autopsy Headrest