Thermo Scientific™ Shandon™ Biopsy Processing/Embedding Cassettes II
Hold biopsy specimens during processing and embedding with the special design of Thermo Scientific™ Shandon™ Biopsy Processing/Embedding Cassettes II.
Manufacturer: Thermo Scientific™ 1001101
 View more versions of this product
Catalog No. 10-011-01
  Check Availability
Add to Cart
    Specifications
    Pore Style Small Square
    Material Acetal polymer
    Writing Face Angle 45°
    Color Blue
    Hold biopsy specimens during processing and embedding with the special design of Thermo Scientific™ Shandon™ Biopsy Processing/Embedding Cassettes II.
    • Acetal polymer construction
    • Locking lid attached by two hinges
    • Cassette writing face is at a 45° angle
    • Pore style is small square
    • Variety of colors available
    Ordering Alerts:
    • Supplied in boxes of 500 cassettes with attached lids
  • Description & Specifications
Related Products
Product Content Feedback