Learn More
Thermo Scientific™ Shandon™ Biopsy Processing/Embedding Cassettes II
Hold biopsy specimens during processing and embedding with the special design of Thermo Scientific™ Shandon™ Biopsy Processing/Embedding Cassettes II.
Manufacturer:  Thermo Scientific™ 1001105
 View more versions of this product
Catalog No. 10-011-05
  Check Availability
Add to cart
Specifications
Description
Hold biopsy specimens during processing and embedding with the special design of Thermo Scientific™ Shandon™ Biopsy Processing/Embedding Cassettes II.
  • Acetal polymer construction
  • Locking lid attached by two hinges
  • Cassette writing face is at a 45° angle
  • Pore style is small square
  • Variety of colors available
Ordering Alerts:
  • Supplied in boxes of 500 cassettes with attached lids
Description & Specifications
Specifications
Small Square
45°
Acetal polymer
Biopsy Cassettes, Small Square Pore
Lilac